Trajan SGE 005000 Microvolume Syringe with Fixed Bevel-Tip Needle and Standard Plunger, 100 uL, 50 mm L, 25 G; 1/Pk
Representative image only.
Trajan SGE 005000 Microvolume Syringe with Fixed Bevel-Tip Needle and Standard Plunger, 100 uL, 50 mm L, 25 G; 1/Pk
Trajan | Mfr: 005000| Item: 1600074
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹6,000.00INR / Each
₹6,000.00INR / Each
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • Syringe TypeManual
  • Syringe Capacity (µL)100
  • Needle Tip StyleBevel
  • Needle IncludedY
  • Needle Gauge25
  • DescriptionTrajan SGE Microvolume Syringe with Fixed BT Needle and Standard Plunger, 100 uL, 50 mm L, 25 G; 1/Pk