Trajan SGE 004810 Gas Tight Syringe, Compatible with CTC PAL Systems, Fixed LC/Blunt-Tip Needle, 50 uL, 22 G; 1/Ea
Representative image only.
Trajan SGE 004810 Gas Tight Syringe, Compatible with CTC PAL Systems, Fixed LC/Blunt-Tip Needle, 50 uL, 22 G; 1/Ea
Trajan | Mfr: 004810| Item: 1600068
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹17,500.00INR / Each
₹17,500.00INR / Each
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • Syringe Capacity (µL)50
  • Needle Gauge22
  • DescriptionTrajan SGE Gas Tight Syringe, Compatible with CTC PAL, Fixed LC Needle, 50 uL, 22 G; 1/Ea