Trajan SGE 003000 Microvolume Syringe with Fixed Needle and Standard Plunger, 25 uL, 50 mm L, 25 G, BT; 1/Pk
Representative image only.
Trajan SGE 003000 Microvolume Syringe with Fixed Needle and Standard Plunger, 25 uL, 50 mm L, 25 G, BT; 1/Pk
Trajan | Mfr: 003000| Item: 1601136
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹4,800.00INR / Each
₹4,800.00INR / Each
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Microvolume Syringe with Fixed Needle and Standard Plunger, 25 uL, 50 mm L, 25 G, BT; 1/Pk