Trajan SGE 002208 Gas Tight Syringe, Fixed BT Needle, 10 uL, 70 mm L, 26 G; 1/Ea
Representative image only.
Trajan SGE 002208 Gas Tight Syringe, Fixed BT Needle, 10 uL, 70 mm L, 26 G; 1/Ea
Trajan | Mfr: 002208| Item: 1599943
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹6,700.00INR / Each
₹6,700.00INR / Each
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • Syringe TypeManual
  • Syringe Capacity (µL)10
  • Needle IncludedY
  • Needle Tip StyleBevel
  • Needle Gauge26
  • DescriptionTrajan SGE Gas Tight Syringe, Fixed BT Needle, 10 uL, 70 mm L, 26 G; 1/Ea