Trajan SGE 002080 Removable Needle - Plunger Protection Syringes, 10 uL, 50 mm L, 26 G, Bevel Tip; 6/Pk
Representative image only.
Trajan SGE 002080 Removable Needle - Plunger Protection Syringes, 10 uL, 50 mm L, 26 G, Bevel Tip; 6/Pk
Trajan | Mfr: 002080| Item: 1599933
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹36,300.00INR / Pkg of 6
₹36,300.00INR / Pkg of 6
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Removable Needle - Plunger Protection Syringes, 10 uL, 50 mm L, 26 G, Bevel Tip; 6/Pk