Trajan SGE 000852 Plunger-in-Needle Syringe with Repeating Adaptor, 5 uL, 70 mm L, 23 G, Cone Tip; 1/Ea
Representative image only.
Trajan SGE 000852 Plunger-in-Needle Syringe with Repeating Adaptor, 5 uL, 70 mm L, 23 G, Cone Tip; 1/Ea
Trajan | Mfr: 000852| Item: 1599901
The choice for sensitivity
₹17,100.00INR / Each
₹17,100.00INR / Each
Product Description
Key Features
  • Lowest syringe carryover
  • Reduced nonspecific sample interaction and increased analyte recovery
  • Choice of surface treatment
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Plunger-in-Needle Syringe with Repeating Adaptor, 5 uL, 70 mm L, 23 G, Cone Tip; 1/Ea