Trajan SGE 000570 Plunger-in-Needle Syringe, 1 uL, 115 mm L, 26 G, Cone Tip; 1/Ea
Representative image only.
Trajan SGE 000570 Plunger-in-Needle Syringe, 1 uL, 115 mm L, 26 G, Cone Tip; 1/Ea
Trajan | Mfr: 000570| Item: 1599889
The choice for sensitivity
₹10,600.00INR / Each
₹10,600.00INR / Each
Product Description
Key Features
  • Lowest syringe carryover
  • Reduced nonspecific sample interaction and increased analyte recovery
  • Choice of surface treatment
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Plunger-in-Needle Syringe, 1 uL, 115 mm L, 26 G, Cone Tip; 1/Ea