Trajan SGE 000300 Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 23 G, Cone Tip; 1/Ea
Representative image only.
Trajan SGE 000300 Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 23 G, Cone Tip; 1/Ea
Trajan | Mfr: 000300| Item: 1599864
The choice for sensitivity
₹13,200.00INR / Each
₹13,200.00INR / Each
Product Description
Key Features
  • Lowest syringe carryover
  • Reduced nonspecific sample interaction and increased analyte recovery
  • Choice of surface treatment
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 23 G, Cone Tip; 1/Ea