Trajan Graphite Short Ferrule With 3/8" Hole For 3/8" Fittings, Pk10
Representative image only.
Trajan Graphite Short Ferrule With 3/8" Hole For 3/8" Fittings, Pk10
Trajan | Mfr: 072615| Item: 1600728
Recommended applications are column to injector and column to FID connections. Unsuitable for connecting columns to mass spectrometers
₹6,700.00INR / Pkg of 10
₹6,700.00INR / Pkg of 10
Product Description
Key Features
  • Easy to use
  • Forms a reliable seal
  • Reusable; soft material; forms a soft grip to the capillary column
  • 6 month warranty
More About this Item
Variety of hole sizes available for 1/16", 1/8" and 1/4" fittings. 450°C temperature limit.
6 Month Warranty
Specifications
  • DescriptionTrajan Graphite short ferrule with 3/8" hole for 3/8" fittings, PK10