Trajan Graphite Reducing Ferrule for 1/16" Fittings, 1 mm Hole, 10/Pk

Representative image only.

Trajan Graphite Reducing Ferrule for 1/16" Fittings, 1 mm Hole, 10/Pk

Trajan | Mfr: 072625| Item: 1600738

Recommended applications are column to injector and column to FID connections. Unsuitable for connecting columns to mass spectrometers

₹5,000.00INR / Pkg of 10

₹5,000.00INR / Pkg of 10
Product Description

Key Features

  • Easy to use
  • Forms a reliable seal
  • Reusable; soft material; forms a soft grip to the capillary column
  • 6 month warranty

More About this Item

Variety of hole sizes available for 1/16", 1/8" and 1/4" fittings. 450°C temperature limit.

6 Month Warranty
Specifications
  • DescriptionTrajan Graphite ferrule with 1.0 mm hole for 1/16" fittings, PK10
  • Warranty6 month
Reviews
Q&A