Trajan Graphite Ferrules, Compatible with PerkinElmer 1/8" Nut ; 10/Pk
Representative image only.
Trajan Graphite Ferrules, Compatible with PerkinElmer 1/8" Nut ; 10/Pk
Trajan | Mfr: 072624| Item: 1600737
Recommended applications are column to injector and column to FID connections. Unsuitable for connecting columns to mass spectrometers
₹5,000.00INR / Pkg of 10
₹5,000.00INR / Pkg of 10
Product Description
Key Features
  • Easy to use
  • Forms a reliable seal
  • Reusable; soft material; forms a soft grip to the capillary column
  • 6 month warranty
More About this Item
Variety of hole sizes available for 1/16", 1/8" and 1/4" fittings. 450°C temperature limit.
6 Month Warranty
Specifications
  • DescriptionTrajan Graphite ferrules with 0.5 mm hole for use with 1/8" nuts on PerkinElmer injectors and detectors, PK10