Trajan CRS Large-Capacity Moisture Trap without Base Plate, 400 mL Volume, 1/4" Brass Fittings
Representative image only.
Trajan CRS Large-Capacity Moisture Trap without Base Plate, 400 mL Volume, 1/4" Brass Fittings
Trajan | Mfr: 202239XL-B| Item: 3465110
Quick and easy install and removal without disconnecting fittings from the line
₹53,300.00INR / Each
₹53,300.00INR / Each
Buy It Together
Current Item
Trajan CRS Large-Capacity Moisture Trap without Base Plate, 400 mL Volume, 1/4" Brass Fittings
Trajan CRS Large-Capacity Moisture Trap without Base Plate, 400 mL Volume, 1/4" Brass Fittings
3465110
₹53,300.00INR / Each
+ + =
Total Price 137400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Suitable for drying laboratory air or central air supplies
More About this Item
Traps remove moisture, oils, and dust from carrier gases. Large 400-mL capacity means fewer refillings are required. Adsorbent materials are indicating desiccant and 5-Å molecular sieve. Desiccant changes color from blue to pink when depleted.
The silicone O-ring seals maintain pressures to 100 psi. Traps feature transparent acrylic housing. Measure 17"L x 2" dia. Choose a trap with base plate for benchtop mounting. Shpg wt 3 lb (1.4 kg).
Specifications
  • Inlet Port1/4" OD
  • Outlet Port1/4" OD
  • DescriptionLarge-capacity moisture traps; 400 mL volume; 1/4" fitting; no base plate