Trajan CRS Carbon Dioxide Gas Trap, 40 L Volume, 1/4" Brass Fittings
Representative image only.
Trajan CRS Carbon Dioxide Gas Trap, 40 L Volume, 1/4" Brass Fittings
Trajan | Mfr: 202352-B| Item: 3465170
Remove carbon dioxide from your gas stream
Call For Price
Call For Price
Product Description
Key Features
  • Trap features 1/4" brass fittings
  • 3 month warranty
More About this Item
Traps remove carbon dioxide from your gas stream. A 500-mL nickel-plated carbon steel cylinder holds packed absorbent (80% emulsive, 320 g; 20% NaOH, 70 g); withstands up to 1000 psi. Include drying stage to prevent moisture from entering system. If downstream moisture is harmful to application, a second drying stage is recommended.
Trap measures 15"L x 2" dia; frits are integral to each fitting. Regeneration or refilling is not recommended. Shpg wt 3 lb (1.4 kg).
3 Month Warranty
Specifications
  • Inlet Port1/4" OD
  • Outlet Port1/4" OD
  • DescriptionCarbon Dioxide Traps; 40 liters volume; 1/4" fittings