Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Compact and reusable—ideal for transportation monitoring
Download data and prepare reports with thorough data analysis
Stores 30,720 temperature and humidity readings with time and date stamp
Only measures 3.25" x 2" x 1.12" and weighs 0.85 ounces
Sticker on the back for quick reference of the logger settings
1 year warranty
More About this Item
This compact, lightweight data logger is reusable and reprogrammable making it ideal for monitoring refrigerated vaccines, chemicals, pharmaceuticals, and other perishable items during transit. Unit comes preprogrammed to record temperature readings every 5 minutes and 33 seconds for 30 days with alarm setpoints at 2°C (low) and 8°C (high). For complete flexibility, the user-defined recording interval may be set based on the number of desired days to log (from 4 hours to 365 days) or the desired time interval (from every 2 seconds to every 24 hours). Once the memory is full the data logger will stop capturing data ensuring no data is lost.
Software is required to program individual units and download and analyze data. One copy of the software may be used for multiple loggers and allows you to update settings including individual unit name/description, time zone settings, daylight savings time on/off, unit of measure (°F or °C), start delay, recording duration/sampling interval, alarm settings, and user stop key enabling. Requires a standard USB to Mini USB converter cable. Order separately.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
Specifications
Number of Channels1
Input TypeTemperature and humidity
Max Temperature (° F)161.6
Min Temperature (° F)-20.2
Max Temperature (° C)72
Min Temperature (° C)-29
Temperature Resolution (° F)0.1
Temperature Accuracy (° F)±0.4 (32 to 50), ±1 (all other ranges)
Temperature Resolution (° C)0.1
Temperature Accuracy (° C)±0.2 (0 to 10), ±0.5 (all other ranges)
Max Humidity RH (RH)100
Min Humidity RH (RH)0
Humidity Resolution (%)0.1
Humidity Accuracy±3 from 25 to 75 %RH; ±5 %RH otherwise
Memory Size30,720 temperature and humidity readings with time and date stamp
Alarm0 to 99 for "ALARM"
Battery Type1 3V CR2450 lithium (included)
Display TypeLCD
DescriptionTemperature and Relative Humidity Data Logger