Traceable Folding Pocket HACCP Food Thermometer
Representative image only.
Traceable Folding Pocket HACCP Food Thermometer
Traceable | Item: 2025034
Take fast and accurate temperature measurements anywhere
₹10,200.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹10,200.00INR / Each
Product Description
Key Features
  • Rugged IP67 rated waterproof case cleans easily under running water
  • Reliable measurements, guaranteed
  • Precalibrated to NIST-traceable standards
  • Hinged design allows probe to fold away and fit into your pocket
  • Narrow probe tip for quicker response time
  • HACCP compliant for food applications
  • 2 year warranty
More About this Item
The tapered 304 stainless steel probe can be used in solids, semisolids, liquids, and gases. A large, backlit display provides great visibility. Fold up 4.5"L (114 mm) probe adjusts from 0 to 180° for best measurement and viewing angle. Features include °F/°C unit selection, data hold, min/max functions, and auto shut-off after 4 minutes of non-use to conserve battery life.
IP 67NIST Traceable CalibrationTwo year warranty
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)544
  • Min Temperature (° C)-50
  • Max Temperature (° C)290
  • Accuracy±1.8°F; ±3.6°F below -4°F; ±1% of reading above 212°F [±1.0°C; ±2.0°F below -20°C; ±1% of reading above 100°C]
  • Resolution0.1°; 1° above 200°F (93°C)
  • IP RatingIP67
  • Low Battery IndicatorYes
  • Display TypeLCD
  • Height (in)6
  • Battery2 AAA (included)
  • DescriptionPrecalibrated Folding Pocket Thermometer