Traceable Digital Stopwatch

Representative image only.

Traceable Digital Stopwatch

Traceable | Mfr: 1030TR| Item: 9876603

Engineered for ease of operation, accuracy, and reliability!

₹10,600.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹10,600.00INR / Each

Buy It Together

Current Item

Traceable Digital Stopwatch
Traceable Digital Stopwatch
9876603

₹10,600.00INR / Each
+
Traceable Digital Contact/Photo Tachometer
2025026

₹28,600.00INR / Each
+ =
Total Price 39200.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Traceable® Certificate, 48"L lanyard, and battery.

Key Features

  • 0.0005% accuracy—over 100 times more accurate than other stopwatches
  • Ideal for timing enzyme tests, viscosity measurements, animal studies, and other timed lab tests
  • Simple design for ease of use
  • Features large digits for easy readability
  • Water-resistant to seal out moisture, dust, and fumes
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory

More About this Item

This precalibrated stopwatch is designed for ease of use and accuracy. Timer has a timing capacity of 9 hours, 59 minutes, 59 seconds, and 99 hundredths with 0.0005% accuracy. Functions include single-event, time-out, time-in, cumulative split, interval split, and continuous roll-over time.

Compact design with raised buttons fits comfortably in your hand. Tough, water-resistant ABS plastic case features tongue and groove craftsmanship, plus an O-ring to seal out moisture, dust, and fumes encountered in harsh lab, plant, or field environments.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

Conformite EuropeenneNIST Traceable Calibration
Specifications
  • Timing Capacity9 hours, 59 minutes, 59 seconds, 99 hundredths
  • Timing ModesSingle-event, time-out/time-in, continuous timing, cumulative split, interval split
  • Count DirectionUp and down
  • Number of Channels1
  • Resolution0.01 sec
  • Accuracy±0.0005%
  • AlarmAudible alarm before zero is reached in countdown mode
  • DisplaySingle-line LCD, 3/8" H
  • CaseABS Plastic; water-resistant
  • CertificationsNIST-traceable
  • Battery Type1 1.5V Silver Oxide (included)
  • Height (InchShort)2 1/2
  • Depth (in)3/4
  • Width (InchShort)3
  • Height (cm)6.4
  • Depth (cm)1.9
  • Width (cm)7.6
  • DescriptionDigital Stopwatch
Reviews
Q&A