Traceable Digital Programmable Plug-In Controller; 115 VAC

Representative image only.

Traceable Digital Programmable Plug-In Controller; 115 VAC

Traceable | Mfr: 5090TR| Item: 0864515

Provides daily and weekly on/off switching

₹7,200.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹7,200.00INR / Each
Product Description

Includes

Traceable® Certification and batteries.

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • AM/PM day programs allow repeating same cycle every day, weekdays only, weekends only or any selected days
  • Battery backup memory saves programs in the event of a power failure
  • 1 year warranty

More About this Item

Plug this controller into any three-prong outlet to turn electrical equipment on or off up to fourteen times a day, seven days a week. This simple-to-use controller can be programmed with 14 different switchings per day with intervals as short as 1 minute or as long as 168 hours. Use the random function to turn the controller on and off at random intervals.

For your convenience, multiple-day switching groups are preprogrammed: Monday-Friday, Monday-Saturday, Saturday-Sunday, and Monday-Sunday. Controller also includes a 24-hour clock and manual program override. Built-in NiCd battery-powered memory retains programs for 10 days in the event of a power failure.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyCanadian Standards AssociationNIST Traceable Calibration
Specifications
  • Timing Capacity7 days
  • Number of Channels1
  • Number of Circuits1
  • Number of Programs14 on/off events per day
  • Resolution1 min
  • Accuracy0.01%
  • DisplayLCD, 0.5" H
  • CertificationsCSA, NRTL-C
  • Control Output120 VAC, 50/60 Hz, 15 A resistive
  • Power (VAC)120;115
  • Power (Hz)50/60
  • Battery Type2 LR44 1.5V button cell batteries
  • Height (InchShort)4
  • Depth (in)2
  • Width (InchShort)3 1/2
  • Height (cm)10.2
  • Depth (cm)5.1
  • Width (cm)8.9
  • DescriptionDigital Programmable Plug-In Controller; 115 VAC
  • Warranty1 year
Reviews
Q&A