Traceable Digital Bottle Thermometer; Liquid Filled Bottle Sensor
Representative image only.
Traceable Digital Bottle Thermometer; Liquid Filled Bottle Sensor
Traceable | Mfr: 4426TR| Item: 9446056
Temperature-buffered sensor minimizes temperature fluctuation from refrigerator door opening
₹9,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹9,700.00INR / Each
Product Description
Includes
Traceable® Certificate, holder/stand, double back tape, hook and loop tape and battery.
Key Features
  • Waterproof
  • Displays high/low readings
  • Bottle base provides stability
  • °C/°F selectable
  • Features a sensor in nontoxic glycol filled bottle
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, and environmental chambers with this temperature-buffered sensor. The probe is sealed in a miniature bottle filled with nontoxic glycol or glass beads. Bottle and solution are designed to insulate the sensor from rapid changes, such as when a refrigerator door is opened.
Easy-to-read digital display shows current temperatures and min/max by monitoring the high and low readings for any time period. The entire unit is waterproof and comes equipped with a stainless-steel sensor that assures long product life. A chemical-resistant ABS plastic holder provides stability when placing the thermometer on a shelf; easily affix to a refrigerator or any wall. Bottle may be removed from holder. Thermometer measures 1-7/8"L x 1"W x 4-5/8"H; base measures 2-1/4"L x 2-1/4"W x 1-1/8"H.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
Conformite EuropeenneNIST Traceable Calibration1 Year Warranty
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-4
  • Max Temperature (° F)122
  • Min Temperature (° C)-20
  • Max Temperature (° C)50
  • Accuracy±0.5°C between 0 to 50°C, otherwise ±1°C
  • Resolution0.1° from 19.9° to 199.9°, 1° otherwise
  • Battery1 silver oxide lithium (included)
  • Probe TypeLiquid Filled Bottle
  • DescriptionDigital Bottle Thermometer; Liquid Filled Bottle Sensor