Traceable Calibrated pH/ORP Meter

Representative image only.

Traceable Calibrated pH/ORP Meter

Traceable | Item: 9876709

Stick-style design for one-handed operation

₹60,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹60,300.00INR / Each

Buy It Together

Current Item

Traceable Calibrated pH/ORP Meter
Traceable Calibrated pH/ORP Meter
9876709

₹60,300.00INR / Each
+ + +
Replacement pH/ORP Probe
9876808

₹17,400.00INR / Each
=
Total Price 152600.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

pH/ORP probe, 6-pack of One-Shot pH calibration buffers, hard-sided carrying case, 2 CR2032 batteries, and NIST-traceable calibration certificate with data.

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 25-point memory storage with time/date stamp
  • IP67-rated for protection against both dust and water
  • 1 year warranty

More About this Item

This handheld pH meter is ideal for wastewater, water quality, and environmental applications. Battery-operated, compact design comes in a convenient kit—everything you need for field use.

The meter itself is pocket-sized for easy one-handed operation. The all-in-one pH/ORP/temperature probe attaches directly to the meter, or can be extended 3.3 ft (1 m) with the use of a cable (98768-07, not included).

Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Meter TypeHandheld
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01 pH units
  • pH Accuracy±0.02 pH units
  • Buffer RecognitionNIST, USA
  • Calibration1-, 2-, or 3-point
  • Max mV (mV)1000
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Min Temperature (° F)32
  • Max Temperature (° F)140
  • Temperature Resolution0.1° from 1 to 100°
  • Temperature Accuracy±1°C
  • Temperature compensationAutomatic
  • Data Logging (points)25 points with date and time stamp
  • WaterproofYes
  • Battery2 CR2032 Lithium (included)
  • Battery life (hr)25
  • Width (in)2 1/2
  • Length (in)10 1/4
  • Height (in)1
  • Diameter (in)1/2
  • NIST Calibration IncludedYes
  • Probe IncludedYes
  • Accessories Includedprobe, probe extension cable, calibration buffers, batteries, and case
  • DescriptionCalibrated pH/ORP Meter
  • Warranty1 Year
Documents & Certificates

Download your certificate of traceability at certificates.traceablelive.com

Reviews
Q&A