Traceable Analog Wall Clock with Temperature and Humidity Dials

Representative image only.

Traceable Analog Wall Clock with Temperature and Humidity Dials

Traceable | Mfr: 1079TR| Item: 9002678

Precise quartz-crystal timing with temperature and humidity displays

₹14,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹14,500.00INR / Each
Product Description

Includes

Traceable® Certificate, wall-mount screw, and battery.

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Crisp reading dials and bright orange sweeping second hand provide superior visibility

More About this Item

With precise quartz-crystal timing and temperature and humidity displays, this clock provides accurate information about ambient conditions. Large 9" (22.9 cm) diameter clock face with black hour and minute hands and bright orange second hand provide superior visibility. The 3" (7.6 cm) diameter temperature and humidity dials measure temperatures from -37 to 47°C (-34 to 116°F) and RH from 8 to 92%.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

NIST Traceable Calibration
Specifications
  • Clock12 hour
  • Resolution1 sec
  • Accuracy0.01%
  • DisplayAnalog
  • CaseABS plastic
  • CertificationsNIST-traceable
  • Max Temperature (° F)116
  • Min Temperature (° F)-34
  • Max Temperature (° C)47
  • Min Temperature (° C)-37
  • Battery Type1 AA (included)
  • Height (InchShort)11
  • Depth (in)1
  • Width (InchShort)12 1/2
  • Height (cm)27.9
  • Depth (cm)2.5
  • Width (cm)31.8
  • Mounting TypeWall
  • DescriptionAnalog Wall Clock with Temperature and Humidity Dials
Documents & Certificates
Reviews
Q&A