Traceable Alarm Timer/Stopwatch
Representative image only.
Traceable Alarm Timer/Stopwatch
Traceable | Mfr: 5020TR| Item: 9022538
Easily program on the 12-button keypad
₹3,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹3,900.00INR / Each
Product Description
Includes
Traceable® Certificate, clip, magnet, flip-open stand, and battery.
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Features: count up/down, time-out, alarm
  • Numbered keys make it a breeze to set countdown time
  • 0.01% accuracy
  • 1 year warranty
More About this Item
Handy timer counts down with an alarm and counts up with a time-out feature. Individually numbered keys make setting time quicker than units with up/down keys. Program it to count down from 99 minutes, 99 seconds to zero in any second increment. In the stopwatch mode, the unit counts up to 59 minutes, 59 seconds, and automatically rolls over to zero and continues timing. Large, 1/2”-high display and vivid LCD digits continuously show time and are readable from 8 feet. Loud, high-decibel alarm automatically silences after 1 minute or may be turned off manually.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Timing Capacity99 minutes, 99 seconds
  • Count DirectionUp and down
  • Number of Channels1
  • Resolution1 second
  • Accuracy±0.01%
  • CertificationsNIST-traceable
  • Battery Type1 silver oxide lithium (included)
  • DescriptionCalibrated Alarm Timer/Stopwatch