TPI Combustible Gas Leak Detector with Alarm

Representative image only.

TPI Combustible Gas Leak Detector with Alarm

TPI | Mfr: HXG-2D| Item: 1010315

Preset alarm level activates audible and visual alarms to warn you of a hazard

₹184,000.00INR / Each

₹184,000.00INR / Each
Product Description

Includes

Soft carrying case, torx screwdriver, wrist strap, and batteries.

Key Features

  • Digital display shows ppm or %LEL readout
  • Readings in ppm auto range to %LEL when the concentration exceeds 990 ppm
  • Ultra-sensitive sensor detects as little as 10 ppm methane
  • Wide-range tick detection — usable in high gas concentrations
  • Preset alarm level of 10% LEL warns you of dangerous gas levels
  • Mute button quiets the alarm when looking for the source of the leak
  • Rugged, intrinsically safe design
  • 3 year warranty
3 Year Warranty
Specifications
  • Gas MeasuredCombustible Gases
  • Range0 to 990 ppm, 0 to 70% LEL autoranging
  • Methane Sensitivity10 ppm
  • Probe16" (41 cm)
  • Response TimeLess than 1 second
  • Battery Type3 C (included)
  • Battery Life50 hours continuous
  • Length (in)10
  • Height (in)1 5/8
  • Width (in)3 1/2
  • Length (cm)25.4
  • Height (cm)4.1
  • Width (cm)8.9
  • DescriptionCombustible Gas Leak Detector with Alarm
  • Warranty3 year
Reviews
Q&A