TPI A773 Combustion Analyzer Sulfur Filter for Oil Applications
Representative image only.
TPI A773 Combustion Analyzer Sulfur Filter for Oil Applications
TPI | Mfr: A773| Item: 1010418
Optimize performance of your air/gas analyzer
₹7,400.00INR / Each
₹7,400.00INR / Each
Product Description
Key Features
  • Sensors
  • Carrying case
  • Software
  • When selecting oil as fuel, be sure to use this optional oil filter or readings could become erratic
  • 1 year warranty
More About this Item
Replacement sensors, carrying cases, and other accessories to customize your sampling and analysis system.
Leak detector sold sold separately.
1 Year Warranty
Specifications
  • Compatible WithTPI Gas Analyzers
  • DescriptionCombustion Analyzer Sulfur Filter for Oil Applications