Tough-Tags 9188-8690 Teeny Labels, White, 0.81" x 0.28"; 1500/RL

Representative image only.

Tough-Tags 9188-8690 Teeny Labels, White, 0.81" x 0.28"; 1500/RL

Tough-Tags | Mfr: 9188-8690| Item: 6220520

₹18,500.00INR / Roll of 1013

₹18,500.00INR / Roll of 1013
Product Description

Key Features

  • 1 year warranty

More About this Item

Precut to fit perfectly around 0.2-mL PCR microtubes.

1 Year Warranty
Specifications
  • ColorWhite
  • Length (in)0.8125
  • Width (in)0.2812
  • DescriptionTeeny Labels, White, 0.81" x 0.28"; 1500/RL
  • Warranty1 year
Documents & Certificates
Reviews
Q&A