Tough-Tags 9188-1002 Small Labels, Green, 1" x 0.5"; 1000/RL

Representative image only.

Tough-Tags 9188-1002 Small Labels, Green, 1" x 0.5"; 1000/RL

Tough-Tags | Mfr: 9188-1002| Item: 6220504

₹18,400.00INR / Roll of 1005

₹18,400.00INR / Roll of 1005
Product Description

Key Features

  • 1 year warranty

More About this Item

Available in a variety of colors to code your different applications.

1 Year Warranty
Specifications
  • ColorGreen
  • Length (in)1
  • Width (in)0.2812
  • DescriptionSmall Labels, Green, 1" x 0.5"; 1000/RL
  • Warranty1 year
Documents & Certificates
Reviews
Q&A