Tough-Tags 9188-1000 Small Labels, White, 1" x 0.5"; 1000/RL

Representative image only.

Tough-Tags 9188-1000 Small Labels, White, 1" x 0.5"; 1000/RL

Tough-Tags | Mfr: 9188-1000| Item: 6220501

₹18,400.00INR / Roll of 1002

₹18,400.00INR / Roll of 1002
Product Description

Key Features

  • 1 year warranty

More About this Item

Available in a variety of colors to code your different applications.

1 Year Warranty
Specifications
  • ColorWhite
  • Width (in)0.5
  • Length (in)1
  • DescriptionSmall Labels, White, 1" x 0.5"; 1000/RL
  • Warranty1 year
Documents & Certificates
Reviews
Q&A