Tough-Tags 9187-1004 Large Labels, Red, 1.25" x 0.5"; 1000/RL

Representative image only.

Tough-Tags 9187-1004 Large Labels, Red, 1.25" x 0.5"; 1000/RL

Tough-Tags | Mfr: 9187-1004| Item: 6220512

₹21,200.00INR / Roll of 1009

₹21,200.00INR / Roll of 1009
Product Description

Key Features

  • 1 year warranty

More About this Item

Available in a variety of colors to code your different applications.

1 Year Warranty
Specifications
  • ColorRed
  • Length (in)1.2812
  • Width (in)0.2812
  • DescriptionLarge Labels, Red, 1.25" x 0.5"; 1000/RL
  • Warranty1 year
Documents & Certificates
Reviews
Q&A