Tough-Spots 9185-0001 Small Labels, Blue, 3/8" Dia; 1000/RL

Representative image only.

Tough-Spots 9185-0001 Small Labels, Blue, 3/8" Dia; 1000/RL

Tough-Spots | Mfr: 9185-0001| Item: 6220704

₹9,000.00INR / Roll of 1000

₹9,000.00INR / Roll of 1000

Buy It Together

Current Item

Tough-Spots 9185-0001 Small Labels, Blue, 3/8" Dia; 1000/RL
Tough-Spots 9185-0001 Small Labels, Blue, 3/8" Dia; 1000/RL
6220704

₹9,000.00INR / Roll of 1000
+ =
Total Price 34800.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Designed for the top of microcentrifuge tubes. Available in a variety of colors to code your different applications.

1 Year Warranty
Specifications
  • ColorBlue
  • Diameter (in)0.375
  • DescriptionSmall Labels, Blue, 3/8" Dia; 1000/RL
  • Warranty1 year
Documents & Certificates
Reviews
Q&A