Torbal Hex Vise Clamp Grip for Test Stand
Representative image only.
Torbal Hex Vise Clamp Grip for Test Stand
Torbal | Mfr: 901104| Item: 2530484
Various grips to suit your application needs
₹70,500.00INR / Each
₹70,500.00INR / Each
Product Description
Key Features
  • 3 years warranty parts and labor
More About this Item
Compatible with the Torbal STV Test Stand.
The chrome-plated hex vise clamp is ideal when performing peel testing, or gripping thin material such as film, paper, or foil. The vise is equipped with an adjustable hex screw and interlocking claps which allow for secure sample griping without damage.
3 Year Warranty
Specifications
  • DescriptionHex Vise Clamp Grip for Test Stand
  • Warranty3 years parts & labor