Tip Biosystems PC-502 Handheld Photometer Cell System Kit, 340, 570, and 600 nm

Representative image only.

Tip Biosystems PC-502 Handheld Photometer Cell System Kit, 340, 570, and 600 nm

| Mfr: 500008-PC| Item: 1601701

Be mobile with this ultrafast portable spectrophotometer

₹279,000.00INR / Case of 1

₹279,000.00INR / Case of 1

Buy It Together

Current Item

Tip Biosystems PC-502 Handheld Photometer Cell System Kit, 340, 570, and 600 nm
Tip Biosystems PC-502 Handheld Photometer Cell System Kit, 340, 570, and 600 nm
1601701

₹279,000.00INR / Case of 1
+ =
Total Price 364400.00 ADD 2 ITEMS TO CART
Product Description

Includes

Device includes: Li-ion rechargeable battery and microUSB charging cable. Kits add on: stand holder, rack of 88 disposable CuveTips, CuveTip rack box, and tip disposal box.

Key Features

  • Instant on-spot quantification of nucleic acids, proteins, cells density and many more
  • Robust optical system with precision ±0.02 AU
  • Geo tagging, photo, notes, date and time embedded in data file and real-time data sharing
  • Wireless connectivity to iOS/Android™ tablets and smartphones via app
  • Preprogrammed applications selectable by user
  • Unique disposable CuveTip® eliminates the need for sample transfer into a cuvette
  • No maintenance or lamp change required
  • 1 year warranty

More About this Item

The Photopette optical system is a multicomponent that integrates a photodiode detector with light emitting diodes (LEDs). It also has a built-in “environmental light correction” and can be used in the lab environment and outdoor without shielding environmental light from the sample. The photometer works together with a smartphone or computing device using a dedicated application which connects via a Bluetooth® link for taking measurements and data transfer.

The dual-purpose disposable reflective CuveTip was designed to act as a sample holder and a light-reflecting system. A minimum volume of 40 μL is required to fill the cavity of the CuveTip; this volume is recovered after the measurement.

Choose from four Photopette models. The Bio model allows direct measurement of DNA/RNA, protein concentration, and cell density. The Cell model permits fast measurement in the cell culture hood. The Turbidity model is ideal for wastewater and environmental monitoring applications. The Custom model lets you specify you three wavelengths.

Applications:

• Absolute quantification of cell density through calibration curve
• Mammalian, bacterial or yeast cell count
• Cell viability test using Resazurin dye
• Enzymatic assays for metabolites
• NAD/NADH based enzymatic assays
• DNA, RNA and Oligo concentration determination
• Bradford assay
• Direct protein concentration
• Lactate, pyruvate, glucose and alcohol

Designed for:
• Academic teaching laboratories (life science)
• Cell culture or tissue culture
• DNA and protein quantification
• Outdoor water and chemical analysis
• Farming and livestock
• Food and beverages

1 Year WarrantyConformite Europeenne
Specifications
  • Min Wavelength (nm)340
  • Max Wavelength (nm)600
  • Wavelength Accuracy±2.0 nm
  • Display TypeLED
  • DescriptionHandheld Photometer Cell System Kit, 340, 570, and 600 nm
  • Warranty1 year
Reviews
Q&A