ThermoSafe 390 Dry Ice Storage Insulated Field Carrier, polyethylene, 1 cu ft
Representative image only.
ThermoSafe 390 Dry Ice Storage Insulated Field Carrier, polyethylene, 1 cu ft
ThermoSafe | Mfr: 390| Item: 0372020
Handle for easy carrying - 1 cu ft capacity
₹219,000.00INR / Each
₹219,000.00INR / Each
Buy It Together
Current Item
ThermoSafe 390 Dry Ice Storage Insulated Field Carrier, polyethylene, 1 cu ft
ThermoSafe 390 Dry Ice Storage Insulated Field Carrier, polyethylene, 1 cu ft
0372020
₹219,000.00INR / Each
+
Tempshield MAM Water-resistant gloves, 15"L, medium, 1 pair
0911312
₹46,100.00INR / Pair of 1
=
Total Price 265100.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
Insulated field carrier has efficient, 2-1/4" thick polyurethane foam insulation core and rugged, double-wall polyethylene construction. Includes carrying handle, hinged lid, and adjustable latches. Inside dimensions are 16-5/8"L x 10-1/4"W x 10-3/4"H.
Specifications
  • Capacity (cu ft)1
  • DescriptionDry Ice Storage Insulated Field Carrier, polyethylene, 1 cu ft