ThermoSafe 3730-20 Compact Insulated Shipper, 3.5 x 3 in, 1/Pk

Representative image only.

ThermoSafe 3730-20 Compact Insulated Shipper, 3.5 x 3 in, 1/Pk

ThermoSafe | Mfr: 3730-20| Item: 0373020

Ship nonhazardous materials

₹6,300.00INR / Each

₹6,300.00INR / Each
Product Description

Key Features

  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.

More About this Item

Compact shippers have a cylindrical interior to hold tubes, vials, and other small items.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)3 1/4
  • Outside Width (in)6
  • Outside Length (in)6
  • Outside Height (in)6
  • Inside Diameter (ID) (in)3
  • Inside Height (cm)8.3
  • Outside Width (cm)15.2
  • Outside Length (cm)15.2
  • Outside Height (cm)15.2
  • Inside Diameter (ID) (cm)7.6
  • Shipping Weight (oz)1.6
  • Shipping Weight (g)45
  • DescriptionCompact Insulated Shipper, 3.5 x 3 in, 1/Pk
Reviews
Q&A