ThermoSafe 358UPS Boston Round Bottle Shipper, Vertical, 16 oz/473 mL; 60/Cs

Representative image only.

ThermoSafe 358UPS Boston Round Bottle Shipper, Vertical, 16 oz/473 mL; 60/Cs

ThermoSafe | Mfr: 358UPS| Item: 0372915

Economical, expanded polystyrene shippers (EPS) for nonhazardous liquids

₹33,300.00INR / Case of 60

₹33,300.00INR / Case of 60
Product Description

More About this Item

A fast, cost-effective way to pack and transport bottles—just tape, label, and ship. Bottles are not included.

Specifications
  • Bottle Capacity (mL)473
  • Inside Height (in)7 5/8
  • Outside Width (in)4 3/4
  • Outside Length (in)4 3/4
  • Outside Height (in)10
  • Inside Diameter (ID) (in)3
  • Inside Height (cm)19.4
  • Outside Width (cm)12.1
  • Outside Length (cm)12.1
  • Outside Height (cm)25.4
  • Inside Diameter (ID) (cm)7.6
  • Shipping Weight (oz)192
  • Shipping Weight (g)5443
  • DescriptionBoston Round Bottle Shipper, Vertical, 16 oz/473 mL; 60/Cs
Reviews
Q&A