Thermo Scientific VERSA STAR pH Module
Representative image only.
Thermo Scientific VERSA STAR pH Module
Thermo Scientific | Mfr: VSTAR-PH| Item: 5882521
Add modules to configure the VERSA STAR meter for what you need now and for later
₹221,000.00INR / Each
₹221,000.00INR / Each
Product Description
Includes
Module for VERSA STAR meter. Purchase electrodes separately.
Key Features
  • Simply click module into back of meter to get started
  • 3 year warranty
More About this Item
Five parameter modules are avialble for the VERSA STAR meter, including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, or add a module to an exising meter package to create your perfect meter.
One or more pH/Temperature modules can be added to any Versa Star meter.
3 Year WarrantyIP 67
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 6 points
  • DescriptionpH Module