Thermo Scientific SureSTART™ Certified Shell Vial and Plug Kit, Level 1, Clear Glass, 1.25 mL; 200/Pk
Representative image only.
Thermo Scientific SureSTART™ Certified Shell Vial and Plug Kit, Level 1, Clear Glass, 1.25 mL; 200/Pk
Thermo Scientific | Mfr: 6EKSHV8-1| Item: 0892247
Vial kits combine vials and plugs for your convenience
₹11,400.00INR / Pkg of 200
₹11,400.00INR / Pkg of 200
Product Description
Includes
200 vials and 200 plugs.
Key Features
  • Certified performance level 1 for everyday analysis and chromatography approved
  • Constructed of glass vials and polyethylene plugs
  • 6 month warranty
More About this Item
Meet your certification and documentation needs with the Level 1 Thermo Scientific™ SureSTART™ specification certified shell vial and plug kits. These convenience kits contain 1.25 mL shell screw vials in clear glass and transparent 8 mm polyethylene plug closures.
Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
6 Month Warranty
Specifications
  • Volume (mL)1.25
  • Tube MaterialGlass
  • Tube ColorClear
  • Cap TypePlug
  • Cap MaterialPolyethylene
  • Cap colorTransparent
  • Level1
  • DescriptionCertified Shell Vial and Plug Kit, Level 1, Clear Glass, 1.25 mL; 200/Pk
  • Warranty6 Months