Thermo Scientific STAR A214 Star A214 pH/ISE Ammonia Meter Kit

Representative image only.

Thermo Scientific STAR A214 Star A214 pH/ISE Ammonia Meter Kit

Thermo Scientific | Mfr: STARA2146| Item: 5882530

Durability and dependability make it the ideal choice for every lab, every sample, every day

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Thermo Scientific STAR A214 Star A214 pH/ISE Ammonia Meter Kit
Thermo Scientific STAR A214 Star A214 pH/ISE Ammonia Meter Kit
5882530

Call For Price
+ + =
Total Price 197700.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Multilanguage interface supports English, Spanish, French, Italian, German, and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separately)
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A214 pH, ISE, mV, RmV and temperature meter features an intuitive user interface with plain language screen prompts. A large backlit, graphic LCD shows measurement with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

Up to five point automatic calibration with USA/NIST and DIN buffers, auto-buffer recognition, and recalibration feature to fix calibration errors. GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

High-performance ammonia ISE (05722-18), ROSS Ultra pH electrode (05718-70), Star ATC probe (58824-48), Star stirring probe (58823-45), 475-mL each of 1000 ppm standard, ISA, and storage solution, swing-arm electrode holder, and universal power adapter.

3 Year WarrantyIP54 RatedISO9001:2000 Certified Supplier
Specifications
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 ppm
  • Max ISE19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.2 or 0.05% of reading
  • Dissolved Oxygen Accuracy±0.5% of reading
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionStar A214 pH/ISE Ammonia Meter Kit
  • Warranty3 year
Reviews
Q&A