Thermo Scientific Smart2Pure Water System 3LPH
Representative image only.
Thermo Scientific Smart2Pure Water System 3LPH
Thermo Scientific | Mfr: 08.2030| Item: 1595462
Ideal solution for low-volume requirements of ASTM Type I and II water
₹482,000.00INR / Each
₹482,000.00INR / Each
Product Description
Key Features
  • Small footprint and attractive design—perfect for any lab
  • 1 year warranty
More About this Item
These complete compact systems produce 3 or 6 liters of ultrapure water per hour. Units have significantly low running costs due to individual replacement of reverse osmosis and ion exchange cartridges.
Built-in 6-L tank is made of ultrapure water-resistant, pigment-free and food-safe polyethylene and has a conical bottom outlet for complete emptying; can be easily cleaned and disinfected. Tank is equipped with a sterile vent filter and a pressure pump for supplying equipment connected downstream (e.g. autoclaves, analyzers).
Systems feature pretreatment module, integrated reverse osmosis module, ultrapure water set, sterilizing filter, and tank with pressure/recirculation pump. Operate on your lab bench or mount directly to the wall. Swivel-mounted display is easy to read from any angle.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionSmart2Pure Water System 3LPH