Thermo Scientific ROSS™ PerpHecT pH Electrode, 3.3-ft (1-m) Cable

Representative image only.

Thermo Scientific ROSS™ PerpHecT pH Electrode, 3.3-ft (1-m) Cable

Thermo Scientific | Mfr: 8220BNWP| Item: 0571821

Easily measure small samples with ROSS precision

₹148,210.00INR / Each

₹148,210.00INR / Each

Buy It Together

Current Item

Thermo Scientific ROSS™ PerpHecT pH Electrode, 3.3-ft (1-m) Cable
Thermo Scientific ROSS™ PerpHecT pH Electrode, 3.3-ft (1-m) Cable
0571821

₹148,210.00INR / Each
+
Thermo Scientific 810007 electrode fill solution, 3 M KCl 5/cs
0573052

₹24,600.00INR / Case of 5
=
Total Price 172810.00 ADD 2 ITEMS TO CART
Product Description

Includes

3.3-ft (1-m) cable, BNC connector, and 50 mL of filling solution.

Key Features

  • Utilizes ROSS sensor technology for optimal temperature response and stability
  • Fits into very small vessels—sensing tip diameter is only 3 mm for samples as small as 15 µL
  • No breaking—PTFE sleeve protects electrode
  • 1 year warranty

More About this Item

This refillable microelectrode features Orion’s renowned ROSS technology for fast, accurate, and stable pH readings. Response time is 0.01 pH units in 30 seconds while drift is less than 0.002 pH units per day. Double-junction system makes this microelectrode compatible with Tris buffers and samples containing proteins and enzymes. Non-mercury electrode is a great alternative to calomel-based microelectrodes.

1 Year WarrantyISO9001:2000 Certified Supplier
Specifications
  • ApplicationsSmall vessel measurements
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionSure-flow
  • Reference ElectrolyteROSS
  • Min pH (pH)0
  • Max pH (pH)14
  • Accuracy±0.01 pH
  • Temperature Accuracy±0.5°C
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Diameter (mm)3
  • Length (mm)155
  • Cable Length (m)1
  • DescriptionPerpHecT pH electrode,3.3-ft (1-m) cable
  • Warranty1 year
Reviews
Q&A