Thermo Scientific Pure water system Pacific UP 3 l/h with ion exchanger filter set and 4 line display
Representative image only.
Thermo Scientific Pure water system Pacific UP 3 l/h with ion exchanger filter set and 4 line display
Thermo Scientific | Mfr: 08.4103| Item: 1595450
Exceeds industry standards with purity up to 15M -cm
Call For Price
Call For Price
Product Description
Key Features
  • Exceeds Type 2 ASTM and CLSI-CLRW standards produced directly from tap water
  • Automatic and economical production of high purity water for daily requirements of 20 to 200 liters/day
  • Microprocessor controlled for automatic operation with permanent monitoring of critical parameters
  • Upgradable systems from 3 to 40 liters/hr
  • Design allows for bench or wall mounting for greater versatility in the lab
  • 1 year warranty
More About this Item
This multistep purification system is ideal for all daily applications requiring pure Type 2 water, such as preparing microbiological media and buffers, glassware washing, and as a feed water source for Type 1 (ultrapure) water systems.
It combines pretreatment, reverse osmosis, high purity water cartridge into a single system that is easy to use and maintain. The unique recirculation feature maintains stored water and eliminates degradation for superior purity.
The flexible control panel can be tilted for keypad operations. Multifunctional graphic display provides real-time information on water quality and other system performance and diagnostics.
Order optional accessories separately.
Conformite Europeenne1 Year Warranty
Specifications
  • DescriptionPure water system Pacific UP 3 l/h with ion exchanger filter set and 4 line display