Thermo Scientific PC 300-S13 12L Heated Bath, Amb +13 to 300C, 208V

Representative image only.

Thermo Scientific PC 300-S13 12L Heated Bath, Amb +13 to 300C, 208V

Thermo Scientific | Mfr: 1591135| Item: 1214826

Superior performance for the most demanding applications

Call For Price

for an additional
₹68,800.00INR / Each
for an additional
₹74,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Thermo Scientific PC 300-S13 12L Heated Bath, Amb +13 to 300C, 208V
Thermo Scientific PC 300-S13 12L Heated Bath, Amb +13 to 300C, 208V
1214826

Call For Price
+ =
Total Price 74800.00 ADD 2 ITEMS TO CART
Product Description

Includes

Plumbing connection for M16x1, ¼” NPT male, 3/8” O.D.

Key Features

  • Ramp programming (PC 200, 5x10 segments; PC 300, 10x10 segments), with edit during ramp feature
  • Large color LCD (262K colors) — graphical display of operating ramp program
  • Pump speed adjustment to 10 levels for turbulence control
  • 3 year warranty

More About this Item

Building on the capabilities of the Standard (SC) and Advanced (AC) controllers in the SAHARA family, the Premium (PC) model encompasses all of the features of the SC and AC baths, but then provides greater control and flexibility. Use the 10-key access keypad for fast and intuitive software navigation. Application identification (Unit control parameters are optimized for the application behavior, so that temperature control can be as fast as possible with zero over swing or under swing). User level management, e.g. for application temperature limitation: set high and low temperature limits to the system not allowing another user to set the temperature higher or lower than is established. Data logging: all ring buffer data will be stored for serial communication. Extra communication option for Profibus, CAN bus Analog I/O. User selectable high temperature cut-off (only PC 300).

UL Listed.3 Year Warranty
Specifications
  • Bath TypeHeating Circulating Bath
  • Reservoir Capacity (L)12
  • Max Temperature (° C)300
  • Min Temperature (° C)Ambient
  • Low-Level CutoffYes
  • High-Temperature CutoffYes
  • Pressure Pump Max Head At 0 Flow (PSI)6.9
  • Power (VAC)230
  • Power (Hz)50/60
  • Description12L Heated Bath, Amb +13 to 300C, 208V
  • Warranty3 year
Reviews
Q&A