Thermo Scientific P9DS-1 Owl Large-Vertical Double-Sided Cell, 16 x 14 cm gel size

Representative image only.

Thermo Scientific P9DS-1 Owl Large-Vertical Double-Sided Cell, 16 x 14 cm gel size

Thermo Scientific | Mfr: P9DS-1| Item: 2855405

Larger-size cassette gives you the maximum number of samples and separation length

₹533,000.00INR / Each

₹533,000.00INR / Each

Buy It Together

Current Item

Thermo Scientific P9DS-1 Owl Large-Vertical Double-Sided Cell, 16 x 14 cm gel size
Thermo Scientific P9DS-1 Owl Large-Vertical Double-Sided Cell, 16 x 14 cm gel size
2855405

₹533,000.00INR / Each
+
Enduro E0160 Gel XL Electrophoresis System 120V
1731411

₹228,000.00INR / Each
+ =
Total Price 768900.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

upper buffer chamber with internal cooling core, lower buffer chamber, safety lid with attached power supply leads, four blank glass plates, four notched glass plates, blocking plate for single gel operation, and 1.5-mm thick spacer set.

Key Features

  • Upper buffer chamber is extended, providing quick and easy gel casts
  • Built-in cooling system circulates cold tap water for fast run times and flat, even banding patterns
  • Simple clamping mechanism for trouble-free loading of glass cassettes and precast gels
  • 3 year warranty

More About this Item


Model 28554-05 also includes two 1.5-mm thick 10-well combs and two 1.5-mm thick 15-well combs.

3 Year Warranty
Specifications
  • Buffer Solution Required (mL)700
  • Gel Thickness (mm)1.5
  • Cell Width (in)5.5
  • Cell Height (in)8.25
  • Cell Depth (in)9.5
  • Cell Width (cm)13.97
  • Cell Height (cm)20.96
  • Cell Depth (cm)24.13
  • Glass Width (in)6.3
  • Glass Length (in)5.51
  • Glass Width (cm)16
  • Glass Length (cm)14
  • DescriptionOwl Large-Vertical Double-Sided Cell, 16 x 14 cm gel size
  • Warranty3 year
Reviews
Q&A