Thermo Scientific Orion Star T900 Series Power Supply, US; 110 VAC

Representative image only.

Thermo Scientific Orion Star T900 Series Power Supply, US; 110 VAC

Thermo Scientific | Mfr: START-PS1| Item: 9410221

₹13,900.00INR / Each

₹13,900.00INR / Each
Product Description

Key Features

  • 115 VAC US power supply

More About this Item

Enhance your titrators with the following accessories.

Specifications
  • DescriptionT900 Series US Power Supply; 110 V
Reviews
Q&A