Thermo Scientific Orion Star T900 Series Plastic Bottle, 1 L; 1/Ea

Representative image only.

Thermo Scientific Orion Star T900 Series Plastic Bottle, 1 L; 1/Ea

Thermo Scientific | Mfr: START-BT1| Item: 9410210

₹10,400.00INR / Each

₹10,400.00INR / Each
Product Description

Key Features

  • Ideally suited for the T910, T920, T930, and T940 titrators
  • 1 mL capacity
  • Shatterproof plastic
  • Available in pack of 12

More About this Item

Enhance your titrators with the following accessories.

Specifications
  • DescriptionT900 Series Plastic Bottle; 1 L
Reviews
Q&A