Thermo Scientific Nunc 240845 Square BioAssay Dishes, Sterile, Non-Treated, 241 x 20 mm; 20/Cs

Representative image only.

Thermo Scientific Nunc 240845 Square BioAssay Dishes, Sterile, Non-Treated, 241 x 20 mm; 20/Cs

Thermo Scientific | Mfr: 240845| Item: 0192901

Bioassay dish is ideal for agar diffusion assays

₹92,700.00INR / Case of 20

₹92,700.00INR / Case of 20
Product Description

Key Features

  • Constructed of optically clear polystyrene
  • Sterilized by irradiation.
  • External dimensions: 241 x 241 x 20mm

More About this Item

Large, square dishes feature flat bases for uniform distribution of media. Ideal for agar diffusion assays or storage use in screening libraries. Dishes feature stacking ridges and include lids.

Specifications
  • Max Working Well Volume (mL)215
  • Well ShapeSquare
  • Culture Area (cm²)478
  • Surface TreatmentNone
  • Lid (Included)Yes
  • SterileYes
  • Height (mm)20
  • DescriptionSquare BioAssay Dishes, Sterile, Non-Treated, 241 x 20 mm; 20/Cs
Reviews
Q&A