Thermo Scientific Nunc 1568005 Economical Fog Testing System
Representative image only.
Thermo Scientific Nunc 1568005 Economical Fog Testing System
Thermo Scientific | Mfr: 156800| Item: 0192872
Non-treated surface offers low cell attachment
₹150,000.00INR / Case of 100
₹150,000.00INR / Case of 100
Product Description
Includes
extra sterile cap for non-treated flask.
Key Features
  • Certified nonpyrogenic and sterile
  • White caps for easy identification
  • The filter cap ensures constant airflow while minimizing chances of contamination
  • EasyFlask™ allows for easy handling and pipetting
More About this Item
These sterile culture flasks are used for unique cell growth applications that do not require a Nunclon™ Delta surface treatment. Untreated surface offers low attachment. Clear, uniform design provides excellent optical quality, making it a suitable choice for microscopy. Flasks have a white cap for easy identification. All models are certified nonpyrogenic and sterile.
TripleFlask™ model 01928-42 offers a unique three-parallel growth surface that provides a total culture area of 500 cm2 - ideal for scale-up.
EasyFlask™ models 01928-70, -72, and -74 feature an ergonomic design requiring only a 1/3 turn to open and close the cap.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Nonpyrogenic
Specifications
  • MaterialPolystyrene
  • Cap TypeFilter
  • Cap MaterialHDPE
  • Neck TypeAngled
  • Working Volume (mL)30
  • Total Volume (mL)260
  • Culture Area (cm²)75
  • DescriptionEasyFlask™ Cell Culture Flask, Angled Neck, 75 cm2; 100/Cs