Thermo Scientific Nunc 154453 Chamber Slide System, Glass, 1-Well; 16/Pk
Representative image only.
Thermo Scientific Nunc 154453 Chamber Slide System, Glass, 1-Well; 16/Pk
Thermo Scientific | Mfr: 154453| Item: 0183800
Ideal for in situ analysis—cells can be cultured, fixed, stained, and viewed on the same microscope slide
₹62,300.00INR / Case of 16
₹62,300.00INR / Case of 16
Product Description
Key Features
  • Glass slides are specially cleaned for optimal cell attachment and growth.
  • Separate the media chamber from the slide using removal tool (included).
  • Biocompatible acrylic adhesive seals the media chamber and slide, preventing well-to-well contamination.
  • Wells feature a hydrophobic border that defines their growth area.
  • Superfrost® writing area allows for easy sample identification.
More About this Item
Easily perform microscopic analysis of cell cultures using these miniature chamber systems that have removable media chambers. This eliminates the need for cell transfers from a vessel to a slide. The systems consist of a lid for aseptic manipulation, removable polystyrene (PS) media chamber, and a standard 25 x 75 mm slide that functions as the growth substrate.
Choose from either cell-culture treated Permanox® plastic or glass (soda lime) slides. Both are ideal for analysis of fluorescent stained cell cultures as they do not interfere with fluorescent labels. All Chamber Slide systems are presterilized.
Glass slides are specially cleaned for optimal cell attachment and growth. Separate the media chamber from the slide using removal tool (included). Biocompatible acrylic adhesive seals the media chamber and slide, preventing well-to-well contamination. Wells feature a hydrophobic border that defines their growth area. Superfrost® writing area allows for easy sample identification.
Sterile
Specifications
  • Number of Wells1
  • Min Working Well Volume (mL)2
  • Max Working Well Volume (mL)4.5
  • Surface TreatmentUntreated
  • MaterialGlass
  • DescriptionChamber Slide System, Glass, 1-Well; 16/Pk