Thermo Scientific Nalgene 6313-0010 Cryoware markers, 4/pack
Representative image only.
Thermo Scientific Nalgene 6313-0010 Cryoware markers, 4/pack
Thermo Scientific | Mfr: 6313-0010| Item: 0996440
Mark vials, and cardboard and plastic cryogenic boxes
₹8,900.00INR / Pkg of 4
₹8,900.00INR / Pkg of 4
Product Description
Key Features
  • Markers will not smudge or fade at temperatures as low as -196°>C
  • Will mark on surfaces that are free of condensation
  • Extra-fine tip markers allow writing on small surfaces such as cryogenic vials and tubes
  • 1 year warranty
More About this Item
Extra-fine tip markers come in a package including one each of four colors (red, blue, green, and black). To change markings, simply remove with alcohol.
1 Year Warranty
Specifications
  • Tip StyleExtra fine
  • Pen ColorAssorted
  • DescriptionCryoware markers, 4/pack