Thermo Scientific Nalgene 5115-0032 Benchtop Cooler, -15C, 32 x 0.5-2.0 mL Tubes
Representative image only.
Thermo Scientific Nalgene 5115-0032 Benchtop Cooler, -15C, 32 x 0.5-2.0 mL Tubes
Thermo Scientific | Mfr: 5115-0032| Item: 4400110
An excellent alternative to messy ice buckets for keeping reagents and enzymes cool
₹151,000.00INR / Each
₹151,000.00INR / Each
Product Description
Key Features
  • Lids feature a gridded write-on surface for sample identification
  • Stackable design saves valuable lab space
More About this Item
A convenient alternative to messy ice buckets. Body is molded from durable polycarbonate and filled with a nontoxic insulating gel; lid is made of clear polycarbonate (models 44001-10 and -25 have gel-filled white polycarbonate lid). Lids feature a gridded write-on surface for sample identification.
Models 44001-00, -10, and -25 include adapters to accommodate 0.5-mL tubes. Models 44001-20 and
-30 hold tubes up to 125-mm tall when lid is latched. All models have a stainless steel handle for a secure hold during transportation. Stackable design saves space.
Specifications
  • Tubes Held32
  • Tube Size0.5 to 2.0 mL
  • Lowest Set Temperature (° C)-15
  • ColorWhite and Blue
  • Height (in)5 3/4
  • Height (cm)14.6
  • DescriptionBenchtop Cooler, -15C, 32 x 0.5-2.0 mL Tubes