Thermo Scientific Nalgene 5050-0001 Polycarbonate storage boxes, 4/pack
Representative image only.
Thermo Scientific Nalgene 5050-0001 Polycarbonate storage boxes, 4/pack
Thermo Scientific | Mfr: 5050-0001| Item: 0675458
Storage boxes can withstand temperatures down to -196°C
₹27,500.00INR / Pkg of 4
₹27,500.00INR / Pkg of 4
Product Description
Key Features
  • 1 year warranty
More About this Item
Storage boxes 06754-57 and -59 hold microcentrifuge tubes. Box 06754-58 holds bags, ampules, bottles, and other containers; withstand temperatures down to -196ºC. Clear top is keyed for easy sample identification. Boxes fit Nalgene CryoBox™ Rack 06756-08, and other standard stainless steel cryogenic storage racks. Autoclavable.
1 Year Warranty
Specifications
  • Tube SizeNot divided
  • Lowest Set Temperature (° C)-196
  • ColorWhite and Clear
  • Height (in)2
  • Height (cm)5.1
  • DescriptionPolycarbonate storage boxes, 4/pack