Thermo Scientific Nalgene 2752-9050 Natural Dropping Bottle, 15 mL

Representative image only.

Thermo Scientific Nalgene 2752-9050 Natural Dropping Bottle, 15 mL

Thermo Scientific | Mfr: 2752-9050| Item: 3452610

Provides accurate and reliable dispensing of reagents

₹17,700.00INR / Pkg of 25

₹17,700.00INR / Pkg of 25
Product Description

Key Features

  • Chemical resistant
  • Leakproof closure
  • Meets USP Class VI and European Pharmacopoeia standards

More About this Item

Polyethylene dropping bottles provide accurate, reliable, and repeatable dispensing of various reagents. Dropper bottles have excellent chemical resistance. Use opaque bottles with UV-light sensitive reagents. Transparent bottles permit easy content identification. Easily identify reagents with colored leakproof polypropylene closures.
Assembled bottles meet USP Class VI and European Pharmacopoeia standards for biotech, diagnostic, and pharmaceutical uses. Each pack includes bottles, tips, and closures for assembling 25 dropping bottles. Tips easily snap into place for a secure and leakproof fit. Five of each color — red, green, blue, white, and yellow — closures are included.

Specifications
  • Capacity (mL)15
  • Capacity (oz)0.5
  • MaterialLow Density Polyethyene (LDPE)
  • Cap MaterialPolypropylene (PP)
  • Bottle ColorTranslucent
  • AutoclavableNo
  • Outside Diameter (OD) (in)1
  • Outside Diameter (OD) (cm)2.5
  • Overall Height (in)3
  • Overall Height (cm)7.6
  • Description15 mL natural dropping bottle
Reviews
Q&A